Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510310.2 | XP_003510358.1 | 2 - 3 | 444 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase Probe spans exons |