Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509554.1 | XP_003509602.1 | 9 - 10 | 1213 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |