Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643653.1 | XP_007641843.1 | 2 - 3 | 72 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |