Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Theg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502549.1 | XP_003502597.1 | 6 - 7 | 744 | 62 | |
XM_003502550.1 | XP_003502598.1 | 5 - 6 | 672 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |