Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509272.2 | XP_003509320.1 | 5 - 6 | 914 | 95 | |
GenBank mRNA | JI891020.1 | - | 2 - 3 | 363 | 95 | |
JP038793.1 | - | 2 - 3 | 363 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |