Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495468.2 | XP_003495516.1 | 5 - 6 | 556 | 64 | |
XM_007641274.1 | XP_007639464.1 | 5 - 6 | 556 | 64 | ||
GenBank mRNA | JI892731.1 | - | 3 - 4 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |