Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhoq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651904.1 | XP_007650094.1 | 3 - 4 | 455 | 76 | |
GenBank mRNA | JI863696.1 | - | 3 - 4 | 455 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |