Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499891.1 | XP_003499939.1 | 1 - 2 | 388 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Probe spans exons |