Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643419.1 | XP_007641609.1 | 1 - 2 | 402 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Nuclear Receptor Probe spans exons |