Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501542.2 | XP_003501590.2 | 7 - 8 | 1703 | 100 | |
XM_007642196.1 | XP_007640386.1 | 7 - 8 | 1730 | 100 | ||
XM_007642198.1 | XP_007640388.1 | 7 - 8 | 1700 | 100 | ||
XM_007642199.1 | XP_007640389.1 | 8 - 9 | 992 | 100 | ||
XM_007642200.1 | XP_007640390.1 | 8 - 9 | 938 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |