Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505639.2 | XP_003505687.1 | 7 - 8 | 584 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |