Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ido2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502986.1 | XP_003503034.1 | 6 - 7 | 548 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |