Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503109.2 | XP_003503157.1 | 7 - 8 | 980 | 65 | |
XM_007643735.1 | XP_007641925.1 | 7 - 8 | 983 | 65 | ||
XM_007643737.1 | XP_007641927.1 | 7 - 8 | 983 | 65 | ||
GenBank mRNA | JI889128.1 | - | 7 - 8 | 960 | 65 | |
JP047060.1 | - | 7 - 8 | 961 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |