Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652819.1 | XP_007651009.1 | 5 - 6 | 1266 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Recommended Assay Probe spans exons |