Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495899.2 | XP_003495947.1 | 1 - 2 | 1432 | 109 | |
XM_003495900.2 | XP_003495948.1 | 1 - 2 | 1432 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |