Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2c2ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003516001.2 | XP_003516049.1 | 3 - 4 | 816 | 63 | |
GenBank mRNA | JP059100.1 | - | 3 - 4 | 444 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |