Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fosl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650039.1 | XP_007648229.1 | 1 - 2 | 355 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |