Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhipl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003494990.2 | XP_003495038.1 | 7 - 8 | 1835 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |