Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497759.2 | XP_003497807.2 | 1 - 2 | 186 | 63 | |
GenBank mRNA | JI871339.1 | - | 1 - 2 | 155 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |