Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647425.1 | XP_007645615.1 | 3 - 4 | 550 | 80 | |
GenBank mRNA | JI891213.1 | - | 2 - 3 | 240 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Recommended Assay Probe spans exons |