Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ropn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495414.1 | XP_003495462.1 | 3 - 4 | 400 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |