Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499949.2 | XP_003499997.2 | 3 - 4 | 932 | 75 | |
XM_003499950.2 | XP_003499998.2 | 3 - 4 | 1031 | 75 | ||
XM_007640696.1 | XP_007638886.1 | 4 - 5 | 1007 | 75 | ||
XM_007640698.1 | XP_007638888.1 | 4 - 5 | 499 | 75 | ||
XM_007640699.1 | XP_007638889.1 | 2 - 3 | 824 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |