Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il9r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652360.1 | XP_007650550.1 | 2 - 3 | 183 | 87 | |
XM_007652362.1 | XP_007650552.1 | 2 - 3 | 183 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Probe spans exons |