Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504574.2 | XP_003504622.1 | 1 - 2 | 373 | 80 | |
GenBank mRNA | JI886786.1 | - | 1 - 2 | 473 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |