Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648815.1 | XP_007647005.1 | 4 - 5 | 2100 | 83 | |
GenBank mRNA | JI865655.1 | - | 2 - 3 | 306 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |