Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649487.1 | XP_007647677.1 | 4 - 5 | 532 | 75 | |
GenBank mRNA | JI882997.1 | - | 2 - 3 | 379 | 75 |