Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neu4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499265.2 | XP_003499313.1 | 1 - 2 | 213 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |