Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kncn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497399.2 | XP_003497447.1 | 2 - 3 | 222 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |