Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496542.2 | XP_003496590.1 | 2 - 3 | 535 | 72 | |
XM_007651121.1 | XP_007649311.1 | 2 - 3 | 535 | 72 | ||
GenBank mRNA | JP051965.1 | - | 2 - 3 | 325 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |