Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504978.2 | XP_003505026.1 | 2 - 3 | 814 | 94 | |
XM_003504979.2 | XP_003505027.1 | 2 - 3 | 1100 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |