Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stra13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496901.2 | XP_003496949.1 | 2 - 3 | 143 | 55 | |
XM_007654213.1 | XP_007652403.1 | 2 - 3 | 346 | 55 | ||
XM_007654214.1 | XP_007652404.1 | 2 - 3 | 346 | 55 | ||
XM_007654219.1 | XP_007652409.1 | 2 - 3 | 137 | 55 | ||
XM_007654221.1 | XP_007652411.1 | 2 - 3 | 127 | 55 | ||
GenBank mRNA | JP052521.1 | - | 2 - 3 | 120 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |