Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503493.2 | XP_003503541.1 | 5 - 6 | 2533 | 72 | |
XM_003503494.2 | XP_003503542.1 | 5 - 6 | 2395 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |