Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503946.1 | XP_003503994.1 | 5 - 6 | 979 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis 3' Most Probe spans exons |