Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507320.2 | XP_003507368.1 | 4 - 5 | 451 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |