Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496578.2 | XP_003496626.1 | 3 - 4 | 466 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |