Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sap30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504319.2 | XP_003504367.1 | 2 - 3 | 741 | 97 | |
GenBank mRNA | JI867149.1 | - | 2 - 3 | 226 | 97 | |
JP052987.1 | - | 2 - 3 | 226 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |