Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653856.1 | XP_007652046.1 | 3 - 4 | 1256 | 102 | |
GenBank mRNA | JI863373.1 | - | 2 - 3 | 788 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |