Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511473.1 | XP_003511521.1 | 2 - 3 | 406 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Inventoried Probe spans exons |