Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512320.1 | XP_003512368.1 | 5 - 6 | 1202 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |