Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496034.1 | XP_003496082.1 | 1 | 1516 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Ion Channel 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |