Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644849.1 | XP_007643039.1 | 4 - 5 | 677 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Detect Genomic DNA Probe spans exons |