Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642432.1 | XP_007640622.1 | 2 - 3 | 590 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |