Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507074.2 | XP_003507122.1 | 6 - 7 | 1037 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |