Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv39h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505103.2 | XP_003505151.1 | 5 - 6 | 1255 | 68 | |
XM_007645636.1 | XP_007643826.1 | 5 - 6 | 1271 | 68 | ||
GenBank mRNA | JI866759.1 | - | 3 - 4 | 324 | 68 | |
JP064041.1 | - | 1 - 2 | 133 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |