Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509959.2 | XP_003510007.1 | 6 - 7 | 1583 | 86 | |
XM_007650622.1 | XP_007648812.1 | 6 - 7 | 1583 | 86 | ||
XM_007650623.1 | XP_007648813.1 | 5 - 6 | 1460 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |