Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpine3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007620487.1 | XP_007618677.1 | 1 - 2 | 267 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |