Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501770.2 | XP_003501818.1 | 1 - 2 | 427 | 99 | |
XM_007642438.1 | XP_007640628.1 | 1 - 2 | 427 | 99 | ||
XM_007642439.1 | XP_007640629.1 | 2 - 3 | 228 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |