Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501870.2 | XP_003501918.1 | 4 - 5 | 557 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel 3' Most Probe spans exons |