Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509769.2 | XP_003509817.1 | 8 - 9 | 2091 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |