Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641687.1 | XP_007639877.1 | 9 - 10 | 1085 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |